4-Nitrobenzyl alcohol, 99%
4-Nitrobenzyl alcohol, 99%
4-Nitrobenzyl alcohol, 99%
Thermo Scientific Chemicals

4-Nitrobenzyl alcohol, 99%

CAS: 619-73-8 | C7H7NO3 | 153.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 128521000
Price (USD)
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS619-73-8
IUPAC Name(4-nitrophenyl)methanol
Molecular FormulaC7H7NO3
InChI KeyJKTYGPATCNUWKN-UHFFFAOYSA-N
SMILESOCC1=CC=C(C=C1)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point92°C to 95°C
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only