Nitro Blue Tetrazolium Chloride monohydrate, 90%
Nitro Blue Tetrazolium Chloride monohydrate, 90%
Nitro Blue Tetrazolium Chloride monohydrate, 90%
Thermo Scientific Chemicals

Nitro Blue Tetrazolium Chloride monohydrate, 90%

CAS: 298-83-9 | C40H30Cl2N10O6 | 817.644 g/mol
Have Questions?
Catalog NumberQuantity
1285900101 g
Catalog number 128590010
Price (USD)
181.65
Special offer
Online exclusive
Ends: 28-Dec-2026
214.00
Save 32.35
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
181.65
Special offer
Online exclusive
Ends: 28-Dec-2026
214.00
Save 32.35
Each
Chemical Identifiers
CAS298-83-9
IUPAC Name2-{3,3'-dimethoxy-4'-[3-(4-nitrophenyl)-5-phenyl-3H-1,2λ⁵,3,4-tetrazol-2-ylium-2-yl]-[1,1'-biphenyl]-4-yl}-3-(4-nitrophenyl)-5-phenyl-3H-1,2λ⁵,3,4-tetrazol-2-ylium dichloride
Molecular FormulaC40H30Cl2N10O6
InChI KeyFSVCQIDHPKZJSO-UHFFFAOYSA-L
SMILES[Cl-].[Cl-].COC1=CC(=CC=C1[N+]1=NC(=NN1C1=CC=C(C=C1)[N+]([O-])=O)C1=CC=CC=C1)C1=CC=C(C(OC)=C1)[N+]1=NC(=NN1C1=CC=C(C=C1)[N+]([O-])=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Loss on drying=<10 % (105°C)
Appearance (Form)Powder
HPLC>=88 %
Sulfated ash=<0.1 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only