4-Nitrocatechol, 97%
4-Nitrocatechol, 97%
4-Nitrocatechol, 97%
Thermo Scientific Chemicals

4-Nitrocatechol, 97%

CAS: 3316-09-4 | C6H5NO4 | 155.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
10 g
50 g
Packaging:
Glass bottle
Catalog number 128600100
Price (USD)
-
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS3316-09-4
IUPAC Name4-nitrobenzene-1,2-diol
Molecular FormulaC6H5NO4
InChI KeyXJNPNXSISMKQEX-UHFFFAOYSA-N
SMILESOC1=CC=C(C=C1O)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Appearance (Color)Yellow to yellow-green to yellow brown
Melting point173°C to 177°C
Infrared spectrumConforms
Titration with NaOH>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only