2-Nitrophenol, 99%
2-Nitrophenol, 99%
2-Nitrophenol, 99%
Thermo Scientific Chemicals

2-Nitrophenol, 99%

CAS: 88-75-5 | C6H5NO3 | 139.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
500 g
1 kg
Packaging:
Glass bottle
Catalog number 128740050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS88-75-5
IUPAC Name2-nitrophenol
Molecular FormulaC6H5NO3
InChI KeyIQUPABOKLQSFBK-UHFFFAOYSA-N
SMILESOC1=CC=CC=C1[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to yellow-green
Appearance (Form)crystalline mass
Melting point43°C to 47°C
Appearance (Form)Powder or crystals and/or chunks or low melting
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only