Stealth RNAi™ siRNA Actin Positive Control (human)

Catalog number: 12935141

Ambion™  Related applications: RNAi | Transfection

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Stealth RNAi™ siRNA Positive Housekeeping Control Duplexes (human) are ideal for use in RNA interference (RNAi) experiments as a control for assessing knockdown and optimizing RNAi experiments. These controls are: –Designed to efficiently knock-down the intended target –Have been bench tested and are supplied in a ready-to-use format
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Control Type: Gene Knockdown Positive Control
Label or Dye: Unconjugated
Product Line: Stealth RNAi™ siRNA
Product Size: 250 µL
RNAi Type: siRNA
Target Gene: Actin
Shipping Condition: Dry Ice

Contents & storage

This individual control duplex is supplied as 250 µl at 20 µM and includes 1 ml of 1X RNA Annealing/Dilution Buffer.  Store at -20°C. Products are guaranteed stable for 6 months when properly stored.

Documents

Manuals & protocols