1,7-Octadiene, 98.5%
1,7-Octadiene, 98.5%
1,7-Octadiene, 98.5%
Thermo Scientific Chemicals

1,7-Octadiene, 98.5%

CAS: 3710-30-3 | C8H14 | 110.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
Catalog number 129355000
Price (USD)
345.65
Online Exclusive
384.00
Save 38.35 (10%)
Each
Quantity:
500 mL
Request bulk or custom format
Price (USD)
345.65
Online Exclusive
384.00
Save 38.35 (10%)
Each
Chemical Identifiers
CAS3710-30-3
IUPAC Nameocta-1,7-diene
Molecular FormulaC8H14
InChI KeyXWJBRBSPAODJER-UHFFFAOYSA-N
SMILESC=CCCCCC=C
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
GC>=98.0 %
Refractive index1.4210 to 1.4230 (20°C, 589 nm)
Appearance (Form)Liquid
Appearance (Color)Clear colorless to light yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only