n-Octane, 99+%, extra pure
n-Octane, 99+%, extra pure
n-Octane, 99+%, extra pure
Thermo Scientific Chemicals

n-Octane, 99+%, extra pure

CAS: 111-65-9 | C8H18 | 114.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 mL
500 mL
2.5 L
25 L
Packaging:
View all
Glass Bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 129370020
Price (USD)
-
Quantity:
2 mL
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS111-65-9
IUPAC Nameoctane
Molecular FormulaC8H18
InChI KeyTVMXDCGIABBOFY-UHFFFAOYSA-N
SMILESCCCCCCCC
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=99.0 %
Density(15°C) 0.706 to 0.710 kg/l
Appearance (Color)Clear colorless
Infrared spectrumConforms
Appearance (Form)Liquid
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only