2-Octanone, 99+%
2-Octanone, 99+%
2-Octanone, 99+%
Thermo Scientific Chemicals

2-Octanone, 99+%

CAS: 111-13-7 | C8H16O | 128.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
500 mL
2.5 L
Packaging:
Glass bottle
Catalog number 129420025
Price (USD)
129.65
Special offer
Online exclusive
Ends: 28-Dec-2026
152.00
Save 22.35
Each
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
129.65
Special offer
Online exclusive
Ends: 28-Dec-2026
152.00
Save 22.35
Each
Chemical Identifiers
CAS111-13-7
IUPAC Nameoctan-2-one
Molecular FormulaC8H16O
InChI KeyZPVFWPFBNIEHGJ-UHFFFAOYSA-N
SMILESCCCCCCC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.4140 to 1.4160 (20°C, 589 nm)
Appearance (Color)Clear colorless to light yellow
GC>=99.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only