Octanoyl chloride, 99%
Octanoyl chloride, 99%
Octanoyl chloride, 99%
Thermo Scientific Chemicals

Octanoyl chloride, 99%

CAS: 111-64-8 | C8H15ClO | 162.66 g/mol
Have Questions?
Catalog NumberQuantityPackaging
129431000100 gGlass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 129431000
Price (USD)
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS111-64-8
IUPAC Nameoctanoyl chloride
Molecular FormulaC8H15ClO
InChI KeyREEZZSHJLXOIHL-UHFFFAOYSA-N
SMILESCCCCCCCC(Cl)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Titration Argentometric>=98.5 %
Acidity=<1 %
Appearance (Form)Clear liquid
Color scale=<80 APHA
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only