Oxindole, 97%, pure
Oxindole, 97%, pure
Oxindole, 97%, pure
Acros Organics

Oxindole, 97%, pure

CAS: 59-48-3 | C8H7NO | 133.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Packaging:
Glass bottle
Catalog number 129671000
also known as 12967
HSN 29339900
Price (INR)
57,790.00
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
57,790.00
Each
Chemical Identifiers
CAS59-48-3
IUPAC Name1H-indol-2-ol
Molecular FormulaC8H7NO
InChI KeyJHFAEUICJHBVHB-UHFFFAOYSA-N
SMILESOC1=CC2=CC=CC=C2N1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point123°C to 128°C
Appearance (Color)Beige to yellow to orange to pink-brown
Infrared spectrumConforms
Appearance (Form)Powder
HPLC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only