Pentacene, 98%
Pentacene, 98%
Pentacene, 98%
Thermo Scientific Chemicals

Pentacene, 98%

CAS: 135-48-8 | C22H14 | 278.35 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mg
1 g
Packaging:
Glass bottle
Catalog number 129791000
Price (USD)
-
Quantity:
100 mg
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS135-48-8
IUPAC Namepentacene
Molecular FormulaC22H14
InChI KeySLIUAWYAILUBJU-UHFFFAOYSA-N
SMILESC1=CC2=CC3=CC4=CC5=CC=CC=C5C=C4C=C3C=C2C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Dark blue to purple to black
Infrared spectrumConforms
Assay>=97.5 %
Appearance (Form)Crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only