Pentaerythritol, 98%
Pentaerythritol, 98%
Pentaerythritol, 98%
Thermo Scientific Chemicals

Pentaerythritol, 98%

CAS: 115-77-5 | C5H12O4 | 136.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
5 kg
Packaging:
View all
Plastic bottle
Catalog number 129870010
Price (USD)
-
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS115-77-5
IUPAC Name2,2-bis(hydroxymethyl)propane-1,3-diol
Molecular FormulaC5H12O4
InChI KeyWXZMFSXDPGVJKK-UHFFFAOYSA-N
SMILESOCC(CO)(CO)CO
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point257°C to 265°C
Appearance (Color)White
Infrared spectrumConforms
GC>=97.5 %
Appearance (Form)Crystalline powder or crystals
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only