3-Pentanol, 98%
3-Pentanol, 98%
3-Pentanol, 98%
Thermo Scientific Chemicals

3-Pentanol, 98%

CAS: 584-02-1 | C5H12O | 88.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 129990250
Price (USD)
43.65
Special offer
Online exclusive
Ends: 28-Dec-2026
51.50
Save 7.85
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
43.65
Special offer
Online exclusive
Ends: 28-Dec-2026
51.50
Save 7.85
Each
Chemical Identifiers
CAS584-02-1
IUPAC Namepentan-3-ol
Molecular FormulaC5H12O
InChI KeyAQIXEPGDORPWBJ-UHFFFAOYSA-N
SMILESCCC(O)CC
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Refractive index1.4090 to 1.4110 (20°C, 589 nm)
Appearance (Color)Clear colorless
GC>=97.5 %
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only