3-Pentanol, 98%
3-Pentanol, 98%
3-Pentanol, 98%
Thermo Scientific Chemicals

3-Pentanol, 98%

CAS: 584-02-1 | C5H12O | 88.15 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
產品號碼 129995000
價格 (TWD)
-
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
化學識別
CAS584-02-1
IUPAC Namepentan-3-ol
Molecular FormulaC5H12O
InChI KeyAQIXEPGDORPWBJ-UHFFFAOYSA-N
SMILESCCC(O)CC
檢視更多
規格Specification Sheet規格表
Infrared spectrumConforms
Refractive index1.4090 to 1.4110 (20°C, 589 nm)
Appearance (Color)Clear colorless
GC>=97.5 %
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only