For superior PCR performance, upgrade to Invitrogen's new Platinum II Taq Hot-start DNA Polymerase.
Platinum™ Hot Start PCR Master Mixes (2X)
Invitrogen™

Platinum™ Hot Start PCR Master Mixes (2X)

Invitrogen Platinum Hot Start PCR Master Mix offers Platinum Taq DNA polymerase in an optimized PCR buffer with magnesium and dNTPs for convenient PCR setup.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorNo. of Reactions
13000013Colorless200 Reactions
13000012Colorless50 Reactions
13000014Colorless1000 Reactions
13001012Green50 Reactions
13001013Green200 Reactions
13001014Green1000 Reactions
Catalog number 13000013
Price (USD)
427.65
Online exclusive
445.00
Save 17.35
Each
Color:
Colorless
No. of Reactions:
200 Reactions
Request bulk or custom format
Price (USD)
427.65
Online exclusive
445.00
Save 17.35
Each
Invitrogen Platinum Hot Start PCR Master Mix, available in colorless and green formats, offers Platinum Taq DNA polymerase in an optimized PCR buffer with magnesium and dNTPs for convenient PCR setup. It retains all the features of Platinum Taq DNA Polymerase: specificity, robustness, and reliability.

Platinum Green Hot Start PCR Master Mix additionally contains a density reagent and two tracking dyes for direct loading of PCR products on gels.

Platinum Hot Start PCR Master Mix provides a ready-to-use mixture of DNA polymerase, salts, magnesium, and dNTPs, thus reducing the number of pipetting steps during PCR reaction setup. It is supplied with a separate vial of Platinum GC Enhancer, formulated for specific amplification and improved yields of GC-rich targets.

Platinum Taq DNA Polymerase is a recombinant Taq DNA polymerase complexed with proprietary antibodies that inhibit enzyme activity at room temperature, reducing nonspecific target amplification at lower temperatures. The antibodies dissociate during the initial PCR denaturation step, and DNA polymerase regains its full activity. Just as with Taq DNA Polymerase, Platinum Taq DNA Polymerase has a non-template-dependent terminal transferase activity that adds a 3' deoxyadenosine to product ends and has a 5' to 3' exonuclease activity.

Features

  • High specificity and increased yields with antibody-mediated hot-start PCR
  • Convenient room temperature reaction setup
  • Versatile formulation for broad range of amplicons
  • Direct gel loading with green PCR master mix

Applications

  • Hot Start PCR
  • Genotyping
  • Gene expression profiling
  • Next-generation sequencing

Notes

  • Platinum II Taq Hot-start DNA Polymerase is recommended superior PCR performance.
  • For applications that require PCR product analysis by absorbance or fluorescence excitation, colorless format is recommended.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
ColorColorless
Fidelity (vs. Taq)1X
FormatTube
Hot StartBuilt-In Hot Start
No. of Reactions200 Reactions
Overhang3'-A
PolymerasePlatinum Taq DNA Polymerase
Product TypeHot Start PCR Master Mix
Quantity200 reactions
Reaction FormatSuperMix or Master Mix
Shipping ConditionDry Ice
Size (Final Product)5 kb or less
Starting MaterialDNA
Concentration2X
For Use With (Application)Hot-start PCR
GC-Rich PCR PerformanceMedium
Label or DyeUnlabeled
Reaction SpeedStandard
Unit SizeEach
Contents & Storage
• Platinum Hot Start PCR 2X Master Mix, 4 x 1.25 mL
• Platinum GC Enhancer, 2 x 1.25 mL
• Nuclease-free water, 4 x 1.25 mL

Store at -20°C in a non-frost-free freezer.

Frequently asked questions (FAQs)

Do you have any recommendations for troubleshooting the amplification of GC-rich or problematic targets with Platinum Hot Start PCR Master Mix (2X) (Cat. Nos. 13000012, 13000013, 13000014)?

We have the following recommendations:
Increase the extension time from 1 min/kb to 1.5 min/kb.
Vary total PCR cycles from 20-40; 35 cycles is typical.
Try denaturing at 95 degrees C for 45 seconds.
GC-rich or problematic targets work better with MgSO4 instead of MgCl2.
For GC-rich templates, test higher annealing temperatures starting with the temperature that is equal to your primer Tm and in increments of 2 degrees C for up to six different temperatures.

Note: Addition of Platinum GC Enhancer solution enhances the amplification of GC-rich and problematic sequences.

What is the final concentration of MgCl2 in the Platinum Hot Start PCR Master Mix (2X) (Cat. Nos. 13000012, 13000013, 13000014)?

The final concentration is 1.5 mM MgCl2.

When should the GC enhancer be used that comes with the Platinum Green Hot Start PCR 2X Master Mix (2X)?

We recommend using the GC enhancer for targets with >65% GC sequences. We recommend that you have a final concentration of 20% of the GC enhancer in the reaction mix. There is enough GC enhancer included in the kit to add it to every PCR reaction.

Do you offer Platinum Taq DNA Polymerase formats that already contain loading dyes in the reaction buffer?

Yes, we offer several Platinum Taq DNA polymerases and their master mixes, supplemented with tracking dyes for direct loading of PCR products on gels:

- Platinum II Hot-Start Green PCR Master Mix (2X)
- Platinum SuperFi Green DNA Polymerase and Platinum SuperFi Green PCR Master Mix
- Platinum Taq Green Hot-Start DNA Polymerase and Platinum Green Hot-Start PCR Master Mix (2X)
- DreamTaq Green Hot-Start DNA Polymerase and DreamTaq Hot-Start Green PCR Master Mix
- Phusion Green Hot-Start II High-Fidelity DNA Polymerase and Phusion Green Hot-Start II High-Fidelity PCR Master Mix.