1-Penten-3-ol, 97%
1-Penten-3-ol, 97%
1-Penten-3-ol, 97%
Thermo Scientific Chemicals

1-Penten-3-ol, 97%

CAS: 616-25-1 | C5H10O | 86.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 130010250
Price (USD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS616-25-1
IUPAC Namepent-1-en-3-ol
Molecular FormulaC5H10O
InChI KeyVHVMXWZXFBOANQ-UHFFFAOYNA-N
SMILESCCC(O)C=C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
GC>=96.0 %
Refractive index1.4220 to 1.4260 (20°C, 589 nm)
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only