Phenazine, 98%
Phenazine, 98%
Phenazine, 98%
Thermo Scientific Chemicals

Phenazine, 98%

CAS: 92-82-0 | C12H8N2 | 180.21 g/mol
Have Questions?
Catalog NumberQuantityPackaging
13015010010 gGlass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 130150100
Price (USD)
64.65
Special offer
Online exclusive
Ends: 28-Dec-2026
75.30
Save 10.65
Each
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
64.65
Special offer
Online exclusive
Ends: 28-Dec-2026
75.30
Save 10.65
Each
Chemical Identifiers
CAS92-82-0
IUPAC Namephenazine
Molecular FormulaC12H8N2
InChI KeyPCNDJXKNXGMECE-UHFFFAOYSA-N
SMILESC1=CC2=NC3=CC=CC=C3N=C2C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
HPLC>=97.5 %
Appearance (Color)Yellow to brown
Melting point173°C to 177°C
Appearance (Form)Crystalline powder or crystals or needles
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only