ESI Probes
Actual product may vary
Thermo Scientific™
ESI Probes
Choose a Thermo Scientific™ Autosampler Sample Probe to suit your matrix and improve elemental analysis results when using automated sample introduction.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberIncludesDiameter (Metric) Inner
1301630Internal STD Probe, orange0.5 mm
1301600Carrier Probe, orange marker0.5 mm
1301640Sample Probe, blue marker0.8 mm
1301650Sample Probe, grey marker1.0 mm
1301750Sample Probe, orange marker0.5 mm
Catalog number 1301630
Price (USD)
660.00
Each
Includes:
Internal STD Probe, orange
Diameter (Metric) Inner:
0.5 mm
Price (USD)
660.00
Each
Customized autosampler accessories ensure more efficient, reliable analysis for your autosampler and trace elemental analyzer. Choose the appropriate ESI probe from our comprehensive range of autosampler sample probes.
Choose an ESI autosampler sample probe from the following options: 
Standard Sample Probe
  • 0.5 mm ID—typically used with ICP-MS systems
  • 0.8 mm ID—typically used with ICP-OES systems
  • 1.0 mm ID—for special applications
Probes for use with ESI FAST and prepFAST Systems
  • 0.5 mm ID with black/black pump tubing attached—for transport of carrier solution
  • 0.5 mm ID with orange/green pump tubing attached—for transport of the internal standard
Specifications
Diameter (Metric) Inner0.5 mm
IncludesInternal STD Probe, orange
For Use With (Equipment)ESI Autosamplers
TypeAutosampler Sample Probe
Unit SizeEach