Phenethyl alcohol, 99%
Phenethyl alcohol, 99%
Phenethyl alcohol, 99%
Thermo Scientific Chemicals

Phenethyl alcohol, 99%

CAS: 60-12-8 | C8H10O | 122.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
250 mL
1 L
2.5 L
10 L
Packaging:
View all
Glass bottle
Catalog number 130180025
Price (USD)
286.65
Online exclusive
318.00
Save 31.35
Each
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
286.65
Online exclusive
318.00
Save 31.35
Each
Chemical Identifiers
CAS60-12-8
IUPAC Name2-phenylethan-1-ol
Molecular FormulaC8H10O
InChI KeyWRMNZCZEMHIOCP-UHFFFAOYSA-N
SMILESOCCC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Refractive index1.5300 to 1.5350 (20°C, 589 nm)
Appearance (Form)Liquid
GC>=98.5 %
Specific gravity(25/25°C) 1.016 to 1.021
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only