RPMI 1640 Medium, powder, HEPES, 1 L - FAQs
RPMI 1640 Medium, powder, HEPES, 1 L - FAQs
View additional product information for RPMI 1640 Medium, powder, HEPES - FAQs (13018015)
3 product FAQs found
How light sensitive is RPMI 1640 media? Should I also be protecting it from LED light?
While we know that different wavelengths of light are worse than others for exposure, we would recommend as a best practice to protect the medium from all forms of light exposure including LEDs, as much as possible to ensure optimal performance, as several components within the medium are light sensitive, such as vitamins.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
What is the density (g/L) for RPMI 1640 Medium?
We have specific gravity information for RPMI 1640 Medium: 1.006 kg/L. In this case, the specific gravity is the same as density as the solvent is water.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
What is the shelf life of my powdered media once I reconstitute it?
Expiration date of most Gibco reconstituted dry format media (AGT or DPM) has not been established; end users should assess performance and stability of this reconstituted media in a system that is relevant to their process. A dry format product (either DPM or AGT) will age upon storage and while we may not be able to detect which component(s) degrade since we can't measure every component, that doesn't mean that the reconstituted liquid will have the same stability as a fresh liquid made by direct weigh methods.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.