BioLite™ Cell Culture Treated Flasks
Thermo Scientific™

BioLite™ Cell Culture Treated Flasks

Thermo Scientific™ BioLite™ Cell Culture Treated Flasks featuring standard cell culture-treated surface, are a limited line of cell culture vessels for a wide range of cell types.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCulture AreaClosure Type
13018925 cm²Vented
13019075 cm²Vented
130191175 cm²Vented
13019225 cm²Plug Seal
13019375 cm²Plug Seal
130194175 cm²Plug Seal
130330225 cm²Vented
130332225 cm²Plug Seal
Catalog number 130189
Price (USD)
476.65
Online exclusive
518.00
Save 41.35
Case of 200
Culture Area:
25 cm²
Closure Type:
Vented
Price (USD)
476.65
Online exclusive
518.00
Save 41.35
Case of 200
Experience optimal performance with wide selections in size, Thermo Scientific™ BioLite™ cell culture treated flasks with filter cap and seal cap options suit a multitude of experimental needs while providing cost efficiency for a wide selection of applications in your cell culture routine.
  • Sterile per ISO 11137, SAL 10-6
  • Non-cytotoxic
  • Non-pyrogenic
  • BSE/TSE Free
  • RNase/DNase Free
  • Human DNA Free
  • USP Class VI raw material
  • Shelf life: 2 years
Specifications
ClassClass VI
Closure TypeVented
No. per Case200/Cs.
No. per Pack5 Pk.
Product TypeCell Culture Flask
Surface CoatingCell Culture Treated
Volume (Metric) Working7 mL
AutoclavableNot Autoclavable
Bottom ShapeFlat
Capacity (Metric)7 mL
Culture Area25 cm²
DisposableSingle-use
GraduatedYes
MaterialPolystyrene
Mouth StyleThreaded
SterilitySterile
Surface TreatmentCell Culture-treated
Unit SizeCase of 200

Frequently asked questions (FAQs)

What is the recommended working volume of Nunc adherent cell culture products?

This depends largely on the cell line and working conditions (considerations include headspace for gas exchange, supplements used, and optimal feeding schedule, among others) but we do offer guidelines for development purposes (0.3 to 0.5 mL per cm2) . Note that these volumes are approximate and may have to be modified for best performance. T25 Flask = 7 mL, T75 Flask = 25 mL, T175 Flask = 55 mL, T225 Flask = 70 mL, 300cm2 Flask = 100 mL, 500 cm2 TripleFlask = 200 mL, 35 mm round culture dish = 3 mL, 60 mm round culture dish = 5 mL, 100 mm round culture dish = 12 mL, 150 mm round culture dish = 35 mL, 4 (rectangular) well Multidish = 5 mL, 8 (rectangular) well Multidish = 3 mL, 4 (round) well Multidish = 1 mL, 6 well Multidish = 3 mL, 12 well Multidish = 2 mL, 24 well Multidish = 1 mL, 48 well Multidish = 0.5 mL.