Bovine Pituitary Extract (BPE)
Gibco™

Bovine Pituitary Extract (BPE)

Bovine Pituitary Extract (BPE)
Have Questions?
Catalog NumberQuantity
1302801425 mg
Catalog number 13028014
Price (USD)
93.50
Each
Quantity:
25 mg
Price (USD)
93.50
Each

The protein concentration of each lot of Bovine Pituitary Extract is determined by the Bradford method. Each vial of BPE contains not less than 25 mg of protein.

For Research Use Only. Not for use in diagnostic procedures.

Specifications
Cell TypeMammalian Cells
Culture TypeMammalian Cell Culture
Quantity25 mg
Shelf Life18 Months
Shipping ConditionDry Ice
FormLiquid
Product TypeBovine Pituitary Extract
pH6 to 8
Unit SizeEach
Contents & Storage
Store in freezer (-5 to -30°C).

Frequently asked questions (FAQs)

Are the Keratinocyte SFM basal medium and supplement kit available separately?

The basal medium is not available for purchase individually, and can only be ordered as part of the Keratinocyte SFM kit (catalog # 17005-042). The supplements are available separately: Bovine Pituitary Extract is catalog # 13028-014 and the EGF is catalog # PHG0311.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Which serum-free media can be used to culture mouse keratinocytes?

The Keratinocyte SFM without calcium chloride (Cat. No. 37010-022) is appropriate for the culture of mouse keratinocytes. The medium should be supplemented with calcium chloride at a final concentration of 0.02 mM.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Which serum-free media can be used to culture human keratinocytes?

There are a variety of options for serum-free culture of human keratinocytes. Keratinocyte SFM (Cat. No. 17005-042) is a widely used kit that includes the basal medium, bovine pituitary extract (BPE) and Epidermal Growth Factor (EGF). If you would like a serum-free, defined culture system, the options are the Defined Keratinocyte SFM kit (Cat. No. 10744-019) or Epilife medium (Cat. No. M-EPI-500-CA) supplemented with EDGS (Cat. No. S-012-5). If you would like an animal origin free defined system, Epilife medium (Cat. No. M-EPI-500-CA) with S7 supplement (Cat. No. S-017-5) is an excellent choice.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Citations & References (1)

Citations & References
Abstract
Laminin alpha 3 LG4 module induces matrix metalloproteinase-1 through mitogen-activated protein kinase signaling.
Authors:Utani A, Momota Y, Endo H, Kasuya Y, Beck K, Suzuki N, Nomizu M, Shinkai H,
Journal:J Biol Chem
PubMed ID:12826666
'The LG4 module of the laminin alpha 3 chain (alpha 3 LG4), a component of epithelial-specific laminin-5, has cell attachment activity and binds syndecan (Utani, A., Nomizu, M., Matsuura, H., Kato, K., Kobayashi, T., Takeda, U., Aota, S., Nielsen, P. K., and Shinkai, H. (2001) J. Biol. Chem. 276, 28779-28788). ... More