trans-2-Phenylcyclopropylamine hydrochloride, 97%
trans-2-Phenylcyclopropylamine hydrochloride, 97%
trans-2-Phenylcyclopropylamine hydrochloride, 97%
Thermo Scientific Chemicals

trans-2-Phenylcyclopropylamine hydrochloride, 97%

CAS: 1986-47-6 | C9H11N·HCl | 169.66 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle
Catalog number 130470010
Price (USD)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS1986-47-6
IUPAC Namehydrogen (1S,2R)-2-phenylcyclopropan-1-amine chloride
Molecular FormulaC9H12ClN
InChI KeyZPEFMSTTZXJOTM-RJUBDTSPSA-N
SMILES[H+].[Cl-].N[C@H]1C[C@@H]1C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder and/or chunks
Titration Argentometric>=96.0 %
Appearance (Color)White to light beige
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only