1-Phenyl-1,2-ethanediol, 97%
1-Phenyl-1,2-ethanediol, 97%
1-Phenyl-1,2-ethanediol, 97%
Thermo Scientific Chemicals

1-Phenyl-1,2-ethanediol, 97%

CAS: 93-56-1 | C8H10O2 | 138.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
View all
Glass bottle
Catalog number 130590050
Price (USD)
30.65
Special offer
Online exclusive
Ends: 28-Dec-2026
36.10
Save 5.45
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
30.65
Special offer
Online exclusive
Ends: 28-Dec-2026
36.10
Save 5.45
Each
Chemical Identifiers
CAS93-56-1
IUPAC Name1-phenylethane-1,2-diol
Molecular FormulaC8H10O2
InChI KeyPWMWNFMRSKOCEY-UHFFFAOYNA-N
SMILESOCC(O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow to light beige
Appearance (Form)Powder or crystalline powder or crystals
Infrared spectrumConforms
Melting point62°C to 68°C
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only