D(-)-alpha-Phenylglycine, 98%
D(-)-alpha-Phenylglycine, 98%
D(-)-alpha-Phenylglycine, 98%
Thermo Scientific Chemicals

D(-)-alpha-Phenylglycine, 98%

CAS: 875-74-1 | C8H9NO2 | 151.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Plastic bottle
Catalog number 130630250
Price (USD)
-
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS875-74-1
IUPAC Name(2R)-2-azaniumyl-2-phenylacetate
Molecular FormulaC8H9NO2
InChI KeyZGUNAGUHMKGQNY-SSDOTTSWSA-N
SMILES[NH3+][C@@H](C([O-])=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Water=<0.5 % (K.F.)
Infrared spectrumConforms
Specific optical rotation-151° to -161° (20°C, 589 nm) (c=5, 1M HCl)
Appearance (Form)Crystalline powder
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only