2-Phenylindole, 98%, Thermo Scientific Chemicals
2-Phenylindole, 98%, Thermo Scientific Chemicals
2-Phenylindole, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Phenylindole, 98%, Thermo Scientific Chemicals

CAS: 948-65-2 | C14H11N | 193.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
Packaging:
View all
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 130680050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS948-65-2
IUPAC Name2-phenyl-1H-indole
Molecular FormulaC14H11N
InChI KeyKLLLJCACIRKBDT-UHFFFAOYSA-N
SMILESN1C2=CC=CC=C2C=C1C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to beige to light green
Appearance (Form)Powder or crystals
Infrared spectrumConforms
Melting point188°C to 191°C
HPLC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only