DL-2-Phenylpropionaldehyde, 98%
DL-2-Phenylpropionaldehyde, 98%
DL-2-Phenylpropionaldehyde, 98%
Thermo Scientific Chemicals

DL-2-Phenylpropionaldehyde, 98%

CAS: 93-53-8 | C9H10O | 134.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Glass bottle
Catalog number 130865000
Price (USD)
-
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS93-53-8
IUPAC Name2-phenylpropanal
Molecular FormulaC9H10O
InChI KeyIQVAERDLDAZARL-UHFFFAOYNA-N
SMILESCC(C=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Refractive index1.5160 to 1.5240 (20°C, 589 nm)
Appearance (Color)Clear colorless
Infrared spectrumConforms
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only