3-Phenyl-1-propylamine, 98%
3-Phenyl-1-propylamine, 98%
3-Phenyl-1-propylamine, 98%
Thermo Scientific Chemicals

3-Phenyl-1-propylamine, 98%

CAS: 2038-57-5 | C9H13N | 135.21 g/mol
Have Questions?
Catalog NumberQuantityPackaging
13088025025 gGlass bottle
Catalog number 130880250
Price (USD)
75.65
Online exclusive
83.50
Save 7.85
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
75.65
Online exclusive
83.50
Save 7.85
Each
Chemical Identifiers
CAS2038-57-5
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=97.5 %
Water=<0.5 % (K.F.)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only