Phenyl sulfoxide, 97%
Phenyl sulfoxide, 97%
Phenyl sulfoxide, 97%
Thermo Scientific Chemicals

Phenyl sulfoxide, 97%

CAS: 945-51-7 | C12H10OS | 202.27 g/mol
数量:
25 g
100 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 130970250
価格(JPY)
-
数量:
25 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS945-51-7
IUPAC Name(benzenesulfinyl)benzene
Molecular FormulaC12H10OS
InChI KeyJJHHIJFTHRNPIK-UHFFFAOYSA-N
SMILESO=S(C1=CC=CC=C1)C1=CC=CC=C1
さらに表示
Water=<0.5 % (K.F.)
Appearance (Color)White to off-white
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point66°C to 71°C
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only