Phthalic acid, 99%
Phthalic acid, 99%
Phthalic acid, 99%
Thermo Scientific Chemicals

Phthalic acid, 99%

CAS: 88-99-3 | C8H6O4 | 166.132 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
2.5 kg
Packaging:
Plastic bottle
Catalog number 131070025
Price (USD)
-
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS88-99-3
IUPAC Namebenzene-1,2-dicarboxylic acid
Molecular FormulaC8H6O4
InChI KeyXNGIFLGASWRNHJ-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=CC=C1C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Iron (Fe)=<50 ppm
Melting point200°C to 205°C (Dec)
Titration with NaOH>=98.5 %
Sulfate (SO4)=<50 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only