1,3-Propanediol, 98%
1,3-Propanediol, 98%
1,3-Propanediol, 98%
Thermo Scientific Chemicals

1,3-Propanediol, 98%

CAS: 504-63-2 | C3H8O2 | 76.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
2.5 L
10 L
Packaging:
View all
Glass bottle
Catalog number 131400025
Price (USD)
573.65
Online exclusive
637.00
Save 63.35
Each
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
573.65
Online exclusive
637.00
Save 63.35
Each
Chemical Identifiers
CAS504-63-2
IUPAC Namepropane-1,3-diol
Molecular FormulaC3H8O2
InChI KeyYPFDHNVEDLHUCE-UHFFFAOYSA-N
SMILESOCCCO
View more
SpecificationsSpecification SheetSpecification Sheet
Color scale=<20 APHA
GC>=97.5 %
Infrared spectrumConforms
Water=<0.2 % (K.F.)
Refractive index1.4380 to 1.4400 (20°C, 589 nm)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only