1,3-Propanediol, 98%
1,3-Propanediol, 98%
1,3-Propanediol, 98%
Thermo Scientific Chemicals

1,3-Propanediol, 98%

CAS: 504-63-2 | C3H8O2 | 76.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
2.5 L
10 L
Packaging:
View all
Glass bottle
Catalog number 131402500
Price (USD)
90.65
Special offer
Online exclusive
Ends: 28-Dec-2026
106.00
Save 15.35
Each
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
90.65
Special offer
Online exclusive
Ends: 28-Dec-2026
106.00
Save 15.35
Each
Chemical Identifiers
CAS504-63-2
IUPAC Namepropane-1,3-diol
Molecular FormulaC3H8O2
InChI KeyYPFDHNVEDLHUCE-UHFFFAOYSA-N
SMILESOCCCO
View more
SpecificationsSpecification SheetSpecification Sheet
Color scale=<20 APHA
GC>=97.5 %
Infrared spectrumConforms
Water=<0.2 % (K.F.)
Refractive index1.4380 to 1.4400 (20°C, 589 nm)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only