Propylene carbonate, 99.5%
Propylene carbonate, 99.5%
Propylene carbonate, 99.5%
Propylene carbonate, 99.5%
Thermo Scientific Chemicals

Propylene carbonate, 99.5%

Propylene carbonate, 99.5%, Quantity: 2.5L, Packaging: Glass bottle, Boiling Point: 240.0 deg.C to 243.0 deg.C, Undesignated, Melting Point: -49.0 deg.C, Molecular Weight: 102.09, Percent Purity: 99.5%, Assay Percent Range: 99.4% min. (GC), CAS: 108-32-7, Density: 1.2040g/mL | CAS: 108-32-7 | C4H6O3 | 102.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
1 L
2.5 L
10 L
Packaging:
View all
Glass bottle
Catalog number 131560025
Price (EUR)
113,65
Online exclusive
126,00
Save 12,35
Each
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
113,65
Online exclusive
126,00
Save 12,35
Each
Chemical Identifiers
CAS108-32-7
IUPAC Name4-methyl-1,3-dioxolan-2-one
Molecular FormulaC4H6O3
InChI KeyRUOJZAUFBMNUDX-UHFFFAOYNA-N
SMILESCC1COC(=O)O1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
Color scale=<40 APHA
Infrared spectrumConforms
GC>=99.4 %
Water=<0.2 %
View more

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

RUO – Research Use Only