Pyridine, 99+%, extra pure
Pyridine, 99+%, extra pure
Pyridine, 99+%, extra pure
Thermo Scientific Chemicals

Pyridine, 99+%, extra pure

CAS: 110-86-1 | C5H5N | 79.102 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
1 L
Packaging:
View all
EcoSafPak™ Glass Bottle
Catalog number 131780010
Price (USD)
107.65
Special offer
Online exclusive
Ends: 28-Dec-2026
126.00
Save 18.35
Each
Quantity:
1 L
Packaging:
EcoSafPak™ Glass Bottle
Request bulk or custom format
Price (USD)
107.65
Special offer
Online exclusive
Ends: 28-Dec-2026
126.00
Save 18.35
Each
Chemical Identifiers
CAS110-86-1
IUPAC Namepyridine
Molecular FormulaC5H5N
InChI KeyJUJWROOIHBZHMG-UHFFFAOYSA-N
SMILESC1=CC=NC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
GC>=99.0 %
Water=<0.1 % (Coulometric)
Residue after evaporation=<0.02 %
Refractive index1.5080 to 1.5110 (20°C, 589 nm)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only