3-Quinuclidinone hydrochloride, 99%, Thermo Scientific Chemicals
3-Quinuclidinone hydrochloride, 99%, Thermo Scientific Chemicals
3-Quinuclidinone hydrochloride, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Quinuclidinone hydrochloride, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
View all
Plastic bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 132261000
Price (USD)
-
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS1193-65-3
IUPAC Name1-azabicyclo[2.2.2]octan-3-one hydrochloridyl
Molecular FormulaC7H11ClNO
InChI KeyIUYBBGKYCRJHHV-UHFFFAOYSA-N
SMILES[Cl].O=C1CN2CCC1CC2
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)White to light yellow
Titration Argentometric98.5 to 101.5 % (On dry substance)
Loss on drying=<1.0 % (100°C, 2 h)
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only