MBP Bovine Protein, Natural
Gibco™
MBP Bovine Protein, Natural
MBP Bovine Protein, Natural
Have Questions?
Catalog NumberQuantity
1322801010 mg
Catalog number 13228010
Price (USD)
266.00
Each
Quantity:
10 mg
Price (USD)
266.00
Each
Myelin Basic Protein (MBP) was purified from bovine brain by a modification of the method of Deibler et al. (1). The protein is supplied lyophilized.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Quantity10 mg
Shipping ConditionApproved for shipment on Wet or Dry Ice
FormPowder
SpeciesBovine
Unit SizeEach
Contents & Storage
Store in freezer (-5 to -30°C).