1300 Series Class II, Type A2 Biological Safety Cabinet Package, 3 ft.. width, 10 in. window, SS package, 120V - FAQs
1300 Series Class II, Type A2 Biological Safety Cabinet Package, 3 ft.. width, 10 in. window, SS package, 120V - FAQs
View additional product information for 1300 Series Class II, Type A2 Biological Safety Cabinet Packages - FAQs (1323TS)
1 product FAQs found
Can the 1300 Series Class II, Type A2 Biological Safety Cabinet (Cat. No. 1323TS) be set up on a benchtop without the stand?
Yes, the 1300 Series Class II, Type A2 Biological Safety Cabinet (Cat. No. 1323TS) can be set up on a benchtop without the stand. You have the option to build your own cabinet by selecting the cabinet-only part numbers and adding factory or field-installed accessories.