Styrene, 99%, extra pure, stabilized
Styrene, 99%, extra pure, stabilized
Styrene, 99%, extra pure, stabilized
Thermo Scientific Chemicals

Styrene, 99%, extra pure, stabilized

CAS: 100-42-5 | C8H8 | 104.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 mL
1 L
2.5 L
25 L
Packaging:
View all
Glass Bottle
Catalog number 132790100
Price (USD)
45.65
Special offer
Online exclusive
Ends: 28-Dec-2026
53.60
Save 7.95
Each
Quantity:
10 mL
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)
45.65
Special offer
Online exclusive
Ends: 28-Dec-2026
53.60
Save 7.95
Each
Chemical Identifiers
CAS100-42-5
IUPAC Nameethenylbenzene
Molecular FormulaC8H8
InChI KeyPPBRXRYQALVLMV-UHFFFAOYSA-N
SMILESC=CC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
Infrared spectrumConforms
Color scale=<20 APHA
Refractive index1.5460 to 1.5480 (20°C, 589 nm)
Stabilizer10 to 20 ppm p-tert-Butylcatechol
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only