Nunc™ Cell Factory™ Shaker GEN 2
Nunc™ Cell Factory™ Shaker GEN 2
Nunc™ Cell Factory™ Shaker GEN 2
Nunc™ Cell Factory™ Shaker GEN 2
Thermo Scientific™
Nunc™ Cell Factory™ Shaker GEN 2
Eliminate the need for manual methods of cell detachment, such as shaking and pounding, with the Thermo Scientific™ Nunc™ Cell Factory™ Shaker System.
Have Questions?
Catalog NumberDescription
132849Cell Factory Shaker GEN 2
Catalog number 132849
Price (USD)
-
Request A Quote
Description:
Cell Factory Shaker GEN 2
Eliminate the need for manual methods of cell detachment, such as shaking and pounding, with the Thermo Scientific™ Nunc™ Cell Factory™ Shaker System. This automated system performs a horizontal, continuous accelerated and decelerated movement to the Cell Factory System.
  • Adjustable frequency of the shaking
  • Designed to easily load and unload Cell Factory carts
  • Stainless steel
Ordering Information:
For use with Nunc Cell Factory Side Loading Rack with Cart, please order the Retrofit Clamping Kit (Cat. No. 120289).
Specifications
Certifications/ComplianceThe Cell Factory Shaker complies with the safety requirements of the following standards and guidelines: Machinery Directive 98/37/EC, Low Voltage Directive 2006/96/EC, EMC Directive 2004/108/EC.
DescriptionCell Factory Shaker GEN 2
Electrical Requirements208/240V 50/60Hz
Frequency50/60Hz
Product TypeShaker GEN 2
Voltage208/240V
MaterialStainless Steel
Unit SizeEach