Sulfur trioxide pyridine complex, technical, 48-50% active SO3
Sulfur trioxide pyridine complex, technical, 48-50% active SO3
Sulfur trioxide pyridine complex, technical, 48-50% active SO3
Thermo Scientific Chemicals

Sulfur trioxide pyridine complex, technical, 48-50% active SO3

CAS: 26412-87-3 | C5H5NO3S | 159.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Catalog number 132870025
Price (USD)
-
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS26412-87-3
IUPAC Namepyridine; sulfonylideneoxidane
Molecular FormulaC5H5NO3S
InChI KeyUDYFLDICVHJSOY-UHFFFAOYSA-N
SMILESO=S(=O)=O.C1=CC=NC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to beige
Assay95 to 100 % (Sulfur trioxide pyridine)
Infrared spectrumConforms
Assay48 to 50 % (Active SO3)
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acid
RUO – Research Use Only