3,5-Dimethoxy-4-hydroxybenzaldehyde, 98%
3,5-Dimethoxy-4-hydroxybenzaldehyde, 98%
3,5-Dimethoxy-4-hydroxybenzaldehyde, 98%
Thermo Scientific Chemicals

3,5-Dimethoxy-4-hydroxybenzaldehyde, 98%

CAS: 134-96-3 | C9H10O4 | 182.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 132881000
Price (USD)
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS134-96-3
IUPAC Name4-hydroxy-3,5-dimethoxybenzaldehyde
Molecular FormulaC9H10O4
InChI KeyKCDXJAYRVLXPFO-UHFFFAOYSA-N
SMILESCOC1=CC(C=O)=CC(OC)=C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder or crystals and/or chunks
Infrared spectrumConforms
Melting point110°C to 114°C
Appearance (Color)Light yellow to beige to green to brown
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only