Nunc™ TripleFlask Treated Cell Culture Flasks
Thermo Scientific™

Nunc™ TripleFlask Treated Cell Culture Flasks

Perform large cell mass applications, such as drug screening and scale up, with these cell-culture treated TripleFlask cell culture flasks.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSurface TreatmentClosure TypeBarcode
132913Cell Culture-treatedFilterNo
132867Cell Culture-treatedSolidNo
132920Cell Culture-treatedFilterYes
132925Nunclon DeltaFilterYes
Catalog number 132913
Price (USD)
690.65
Online exclusive
832.00
Save 141.35
Case of 32
Surface Treatment:
Cell Culture-treated
Closure Type:
Filter
Barcode:
No
Price (USD)
690.65
Online exclusive
832.00
Save 141.35
Case of 32

Perform cell culture expansion and experience 500 cm2 total growth area within the footprint of a standard flask using Thermo Scientific Nunc TripleFlask Treated Cell Culture Flasks. Total culture area is achieved by way of the three parallel layers and a straight neck design that works effectively within automation systems. These flasks are subjected to Nunclon Delta treatment, a proprietary cell culture surface treatment that offers maximum adhesion for a broad range of cell types.

Three layers of growth surface provide a total culture area of 500 cm2 in the same footprint as a standard Nunc 175 cm2 flask.

  • Recommended working volume is 200 mL
  • Promotes monolayer adherent cell culture
  • Also available barcoded with large Code 128
  • Nunclon Delta Certificate included in each case for monolayer formation, cloning efficiency, non-cytotoxic, non-pyrogenic, and sterility

Warranty: 90 days

WARNING: Cancer – www.P65Warnings.ca.gov
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
BarcodeNo
Closure TypeFilter
DescriptionCell Culture Treated TripleFlasks
Neck StyleStraight
No. per Case32/Cs.
No. per Pack1 Ea.
Product TypeFlask
ShapeSame external dimensions as the standard Nunc T175 with straight neck
Volume (Metric) Working200 mL
Bottom ShapeFlat
Capacity (Metric)500 cm²
Culture Area500 cm²
DisposableSingle-use
GraduatedYes
MaterialPolystyrene
SterilitySterile
Surface TreatmentCell Culture-treated
Unit SizeCase of 32

Frequently asked questions (FAQs)

What is the difference between the Nunclon Delta surface and a tissue culture (TC) surface from a competitor?

Our Nunclon Delta surface for adherent cells is a standard tissue culture (TC) surface modification that makes the polystyrene surface more hydrophilic. This facilitates maximum adhesion for a broad range of cell types.

The Nunclon Delta surface is Thermo Scientific Nunclon Delta-certified, which is a Thermo Fisher proprietary certification that has a quality standard to help ensure consistency. Every lot of Thermo Scientific Nunclon Delta-certified cell cultureware is tested with Gibco media for:
- Less than 10% growth variation in L929, Hel299 and primary chick embryo cells
- Less than 15% variation colony formation of V79-4 cells

More information and all applicable products can be found in the "Product selection guide for every step of your cell culture workflow" brochure .

Can I use a Nunc Cell Scraper with a Nunc TripleFlask that has Nunclon Delta surface treatment?

The design of the Nunc TripleFlask does not allow for mechanical scraping. However, cells are easily harvested using protein digesting enzymes, such as trypsin, collagenase, and Pronase or by using non-enzymatic dissociation solutions.
The following is a document link regarding culturing techniques for Nunc TripleFlask:
https://assets.thermofisher.com/TFS-Assets/BID/Technical-Notes/culturing-technique-nunc-tripleflask-technical-note.pdf

What is the recommended working volume of Nunc adherent cell culture products?

This depends largely on the cell line and working conditions (considerations include headspace for gas exchange, supplements used, and optimal feeding schedule, among others) but we do offer guidelines for development purposes (0.3 to 0.5 mL per cm2) . Note that these volumes are approximate and may have to be modified for best performance. T25 Flask = 7 mL, T75 Flask = 25 mL, T175 Flask = 55 mL, T225 Flask = 70 mL, 300cm2 Flask = 100 mL, 500 cm2 TripleFlask = 200 mL, 35 mm round culture dish = 3 mL, 60 mm round culture dish = 5 mL, 100 mm round culture dish = 12 mL, 150 mm round culture dish = 35 mL, 4 (rectangular) well Multidish = 5 mL, 8 (rectangular) well Multidish = 3 mL, 4 (round) well Multidish = 1 mL, 6 well Multidish = 3 mL, 12 well Multidish = 2 mL, 24 well Multidish = 1 mL, 48 well Multidish = 0.5 mL.

What is Nunclon Delta?

Nunclon Delta refers to our performance-certified surface for culture of adherent cells, available in a variety of common formats (flasks, plates, multiwell dishes). It is tested with at least four cell lines (varies by product and manufacturing location) commonly used in research, and it should work well for most robust cell lines. It does not involve addition of coating reagent, but rather is manufactured by exposure of the molded products to a proprietary energy source, which renders the surface more hydrophilic by means of increasing the concentration of oxygen-containing (e.g. hydroxyl) molecular groups. It is by this hydrophilic property that anchorage-dependent cells are able to attach to the surface and become confluent.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.