ESI DX Autosamplers
Actual product may vary
ESI DX Autosamplers
ESI DX Autosamplers
ESI DX Autosamplers
ESI DX Autosamplers
Thermo Scientific™
ESI DX Autosamplers
Increase your sample throughput with this selection of advanced autosamplers, compatible with the Thermo Scientific™ high-performance trace elemental analyzers.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
1329530ESI SC-8 DX no FAST
1238540ESI SC-14 DX no FAST
1238570ESI SC-4 DX no FAST
1238590ESI SC-2 DX no FAST
Catalog number 1329530
Price (USD)
-
Type:
ESI SC-8 DX no FAST
Accelerate your sample workflow for trace elemental analysis and free your staff to perform other laboratory tasks with the Elemental Scientific™ (ESI) DX family of autosamplers. These autosamplers feature chemically inert construction and deliver contamination-free sampling for Thermo Scientific™ ICP-OES and ICP-MS instruments. They are precise and durable with flexible configurations to support almost any sample size, from microplates to large bottles.
Launch the automation of your trace elemental analysis laboratory with the most complete range of autosamplers for ICP and ICP-MS. We offer autosampler models with sample rack capabilities from 2 to 14 racks to support your sampling needs. The autosamplers offer fully integrated control using Thermo Scientific™ Qtegra™ Intelligent Scientific Data Solution™ (ISDS) software or ESI software.
Compatible with:
  • Thermo Scientific™ iCAP™ 6000/7000/PRO Series ICP-OES Systems
  • Thermo Scientific™ iCAP™ Q/RQ/TQ Series ICP-MS Systems
  • Thermo Scientific™ Element Series HR-ICP-MS Systems (SC-2DX, SC-4DX and SC-14DX only) with ESI SC software
ESI DX autosamplers deliver:
  • Exceptionally long lifetime, due to durable, dual-rail system
  • Exclusion of metal components from samples using the AutoAlign Arm with fluoropolymer probe guide
  • Reduction in analyte carryover using standard dual flowing rinse stations
  • A reset probe to prevent probe damage
  • Flexible rack configurations, from microplates to large bottles
  • Ability to upgrade to advanced automation with FAST or prepFAST
ESI DX Autosampler Models
  • 2DX Autosampler—2 large racks, 4 microplates
  • 4DX Autosampler—4 large racks, 6 microplates
  • 8DX Autosampler—8 large racks, supplied with Mobile Station
  • 14DX Autosampler—14 large racks, supplied with Mobile Station
Specifications
For Use With (Equipment)iCAP Series ICP-OES, iCAP Q/Qnova Series ICP-MS
TypeESI SC-8 DX no FAST
Unit SizeEach