Squaric Acid Dibutyl Ester, 97%
Squaric Acid Dibutyl Ester, 97%
Squaric Acid Dibutyl Ester, 97%
Thermo Scientific Chemicals

Squaric Acid Dibutyl Ester, 97%

CAS: 2892-62-8 | C12H18O4 | 226.27 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Catalog number 133170010
Price (USD)
36.65
Special offer
Online exclusive
Ends: 28-Dec-2026
43.00
Save 6.35
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
36.65
Special offer
Online exclusive
Ends: 28-Dec-2026
43.00
Save 6.35
Each
Chemical Identifiers
CAS2892-62-8
IUPAC Namedibutoxycyclobut-3-ene-1,2-dione
Molecular FormulaC12H18O4
InChI KeyXBRWELTXMQSEIN-UHFFFAOYSA-N
SMILESCCCCOC1=C(OCCCC)C(=O)C1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Liquid
Refractive index1.4930 to 1.4950 (20°C, 589 nm)
Appearance (Color)Clear colorless to yellow
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only