N-Formylmorpholine, 99%
N-Formylmorpholine, 99%
N-Formylmorpholine, 99%
Thermo Scientific Chemicals

N-Formylmorpholine, 99%

CAS: 4394-85-8 | C5H9NO2 | 115.13 g/mol
Have Questions?
Catalog NumberQuantityPackaging
133805000500 mLGlass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 133805000
Price (USD)
-
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS4394-85-8
IUPAC Namemorpholine-4-carbaldehyde
Molecular FormulaC5H9NO2
InChI KeyLCEDQNDDFOCWGG-UHFFFAOYSA-N
SMILESO=CN1CCOCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Refractive index1.4850 to 1.4880 (20°C, 589 nm)
Infrared spectrumConforms
GC>=98.5 %
Appearance (Color)Clear colorless to yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only