Potassium hydroxide, ca. 85%, for analysis, pellets
Potassium hydroxide, ca. 85%, for analysis, pellets
Potassium hydroxide, ca. 85%, for analysis, pellets
Thermo Scientific Chemicals

Potassium hydroxide, ca. 85%, for analysis, pellets

CAS: 1310-58-3 | HKO | 56.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
2.5 kg
10 kg
25 kg
Packaging:
View all
Plastic bottle
Catalog number 134060010
Price (USD)
82.65
Special offer
Online exclusive
Ends: 28-Dec-2026
97.30
Save 14.65
Each
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
82.65
Special offer
Online exclusive
Ends: 28-Dec-2026
97.30
Save 14.65
Each
Chemical Identifiers
CAS1310-58-3
IUPAC Namepotassium hydroxide
Molecular FormulaHKO
InChI KeyKWYUFKZDYYNOTN-UHFFFAOYSA-M
SMILES[OH-].[K+]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Pellets
Titration with HCl>=85 %
Potassium carbonate=<1 % (K2CO3)
Chloride (Cl)=<70 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only