Activated charcoal, NORIT™ SA 2
Activated charcoal, NORIT™ SA 2
Activated charcoal, NORIT™ SA 2
Thermo Scientific Chemicals

Activated charcoal, NORIT™ SA 2

CAS: 7440-44-0 | C | 12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
2.5 kg
25 kg
Packaging:
View all
Plastic drum
Catalog number 134370025
Price (USD)
213.65
Special offer
Online exclusive
Ends: 28-Dec-2026
251.00
Save 37.35
Each
Quantity:
2.5 kg
Packaging:
Plastic drum
Request bulk or custom format
Price (USD)
213.65
Special offer
Online exclusive
Ends: 28-Dec-2026
251.00
Save 37.35
Each
Chemical Identifiers
CAS7440-44-0
IUPAC Namecarbon
Molecular FormulaC
InChI KeyOKTJSMMVPCPJKN-UHFFFAOYSA-N
SMILES[C]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Black
Loss on drying=<10 %
Appearance (Form)Powder
Molasses number=<390
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Filter aid
RUO – Research Use Only