2-chloro-N-(2,6-dimethylphenyl)acetamide, 98%
2-chloro-N-(2,6-dimethylphenyl)acetamide, 98%
2-chloro-N-(2,6-dimethylphenyl)acetamide, 98%
Thermo Scientific Chemicals

2-chloro-N-(2,6-dimethylphenyl)acetamide, 98%

CAS: 1131-01-7 | C10H12ClNO | 197.66 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Packaging:
Glass bottle
Catalog number 135440100
Price (USD)
71.65
Online exclusive
79.00
Save 7.35
Each
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
71.65
Online exclusive
79.00
Save 7.35
Each
Chemical Identifiers
CAS1131-01-7
IUPAC Name2-chloro-N-(2,6-dimethylphenyl)acetamide
Molecular FormulaC10H12ClNO
InChI KeyFPQQSNUTBWFFLB-UHFFFAOYSA-N
SMILESCC1=CC=CC(C)=C1NC(=O)CCl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to brown
Appearance (Form)Powder or crystalline powder or needles
Infrared spectrumConforms
Melting point143°C to 150°C
HPLC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only