Nunc™ MicroWell™ 96-Well, Nunclon Delta-Treated, Flat-Bottom Microplate
Nunc™ MicroWell™ 96-Well, Nunclon Delta-Treated, Flat-Bottom Microplate
Nunc™ MicroWell™ 96-Well, Nunclon Delta-Treated, Flat-Bottom Microplate
Thermo Scientific™

Nunc™ MicroWell™ 96-Well, Nunclon Delta-Treated, Flat-Bottom Microplate

Color: White; Sterility: Sterile; Lid: With Lid
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNo. of WellsNo. per PackNo. per Case
1361029610 Microplates160 Microplates
136101961 Microplate50 Microplates
Catalog number 136102
Price (USD)
1,219.65
Online exclusive
1,284.00
Save 64.35
Case of 160
No. of Wells:
96
No. per Pack:
10 Microplates
No. per Case:
160 Microplates
Price (USD)
1,219.65
Online exclusive
1,284.00
Save 64.35
Case of 160

Ensure consistent and reliable results using the high quality Thermo Scientific™ Nunc™ F96 MicroWell™ White Polystyrene Plate. White plates are optimized for robotics, plate readers and liquid handling systems and designed for key imaging characteristics.

  • Background, autoflorescence and crosstalk are improved when you use Nunc White microplates.
  • White microplates are recommended for luminescence measurements; with maximum reflection and minimal autoluminescence
  • Working volume range: 50 to 250μL/well
  • Choice of Thermo Scientific™ Nunclon™ Delta surface for cell-based assays
  • Customized bar coding is available
WARNING: Cancer – www.P65Warnings.ca.gov
Specifications
Certifications/ComplianceAll microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004)
CertifiedNunclon Delta certified for monolayer formation, cloning efficiency, non-cytotoxic, non-pyrogenic, and sterility
LidWith Lid
No. per Case160 Microplates
No. per Pack10 Microplates
Packaging10 Pk., 160 Cases
Product TypeMicroplate
Skirt StyleLow Flange Pinchbar
Volume (Metric) Well400 μL
Volume (Metric) Working50 to 200 μL
Array8 x 12
BarcodeCustom Barcoding Available
ColorWhite
MaterialPolystyrene
No. of Wells96
Product LineNunc
SterilitySterile
Surface TreatmentNunclon Delta
Well ShapeFlat Bottom (F-well)
Unit SizeCase of 160

Frequently asked questions (FAQs)

What is the difference between the Nunclon Delta surface and a tissue culture (TC) surface from a competitor?

Our Nunclon Delta surface for adherent cells is a standard tissue culture (TC) surface modification that makes the polystyrene surface more hydrophilic. This facilitates maximum adhesion for a broad range of cell types.

The Nunclon Delta surface is Thermo Scientific Nunclon Delta-certified, which is a Thermo Fisher proprietary certification that has a quality standard to help ensure consistency. Every lot of Thermo Scientific Nunclon Delta-certified cell cultureware is tested with Gibco media for:
- Less than 10% growth variation in L929, Hel299 and primary chick embryo cells
- Less than 15% variation colony formation of V79-4 cells

More information and all applicable products can be found in the "Product selection guide for every step of your cell culture workflow" brochure .

How does cell culturing on Nunc 384-Well Microplates that have a cell culture-treated surface compare with 96-Well Microplates that have a cell culture-treated surface?

The cell culture-treated surfaces are very similar but the wells of the Nunc 384-Well Microplates are smaller than the wells of the Nunc 96-Well Microplate.
Culture recommendations for the Nunc 384-Well Microplates are:
- 15 - 50 µL media per well
- 0.5 - 2.0 x 10^4 cells per well