4-(1-Pyrrolidinyl)piperidine, 99%
4-(1-Pyrrolidinyl)piperidine, 99%
4-(1-Pyrrolidinyl)piperidine, 99%
Thermo Scientific Chemicals

4-(1-Pyrrolidinyl)piperidine, 99%

CAS: 5004-07-9 | C9H18N2 | 154.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 137010010
Price (USD)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS5004-07-9
IUPAC Name4-(pyrrolidin-1-yl)piperidine
Molecular FormulaC9H18N2
InChI KeySTWODXDTKGTVCJ-UHFFFAOYSA-N
SMILESC1CCN(C1)C1CCNCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Crystals
Infrared spectrumConforms
Melting point52°C to 57°C
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only